Learn More
Biomatik Corporation ABHD7 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50009
Encompass_Preferred
ABHD7 Colorimetric Cell-Based ELISA Kit for detecting ABHD7 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8IUS5. Gene ID: 253152. Target Synonyms: Abhydrolase domain-containing protein 7; Epoxide hydrolase-related protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6762