Learn More
Biomatik Corporation Akt2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50024
Encompass_Preferred
Akt2 Colorimetric Cell-Based ELISA Kit for detecting Akt2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P31751. Gene ID: 208. Target Synonyms: kinase Akt2; PKB beta; Protein kinase Akt-2; Protein kinase B; beta; RAC-beta serine/threonine protein kinase; RAC-PK-beta; v-akt murine thymoma viral oncogene homolog 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6774