Learn More
Biomatik Corporation BTK Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50070
Encompass_Preferred
BTK Colorimetric Cell-Based ELISA Kit for detecting BTK in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q06187. Gene ID: 695. Target Synonyms: AGMX1; ATK; Agammaglobulinaemia tyrosine kinase; B cell progenitor kinase; BPK; Bruton's tyrosine kinase; Kinase EMB; kinase Btk. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6811