Learn More
Biomatik Corporation Cadherin-pan Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50074
Encompass_Preferred
Cadherin-pan Colorimetric Cell-Based ELISA Kit for detecting Cadherin-pan in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P12830; P19022; P22223; P55283. Gene ID: 999; 1000; 1001; 1002. Target Synonyms: Cadherin-1; CAM 120/80; Epithelial cadherin; E-cadherin; Uvomorulin; CD324; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3 CDH1; CDHE; UVO. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6815