Learn More
Biomatik Corporation Collagen IV α2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50135
Encompass_Preferred
Collagen IV α2 Colorimetric Cell-Based ELISA Kit for detecting Collagen IV α2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08572. Gene ID: 1284. Target Synonyms: Collagen alpha-2(IV) chain; Canstatin; COL4A2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6861