Learn More
Biomatik Corporation Cyclin E2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50164
Encompass_Preferred
Cyclin E2 Colorimetric Cell-Based ELISA Kit for detecting Cyclin E2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O96020. Gene ID: 9134. Target Synonyms: CCNE2; CGE2; CYCE2; G1/S-Specific cyclin E2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6887