Learn More
Biomatik Corporation DJ-1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50178
Encompass_Preferred
DJ-1 Colorimetric Cell-Based ELISA Kit for detecting DJ-1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q99497. Gene ID: 11315. Target Synonyms: Protein DJ-1; Oncogene DJ1; Parkinson disease protein 7; PARK7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6901