Learn More
Biomatik Corporation EFNB1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50191
Encompass_Preferred
EFNB1 Colorimetric Cell-Based ELISA Kit for detecting EFNB1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P98172. Gene ID: 1947. Target Synonyms: CEK5 ligand; CEK5 receptor ligand; CEK5-L; CEL5-L; EFL-3; EFNB1; ELK ligand; ELK-L; EPH-related receptor tyrosine kinase ligand 2; EPL2; EPLG2; Ephrin-B1 precursor; LERK-2; LERK2; STRA1; STRA1 protein; kinase ephrin-B1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6913