Learn More
Biomatik Corporation eIF4B Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50195
Encompass_Preferred
eIF4B Colorimetric Cell-Based ELISA Kit for detecting eIF4B in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P23588. Gene ID: 1975. Target Synonyms: Eukaryotic translation initiation factor 4B; IF4B; eIF-4B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6916