Learn More
Biomatik Corporation EPHA1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50198
Encompass_Preferred
EPHA1 Colorimetric Cell-Based ELISA Kit for detecting EPHA1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P21709. Gene ID: 2041. Target Synonyms: EPA1; EPH; EPHT; EPHT1; Tyrosine-protein kinase receptor EPH. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6919