Learn More
Biomatik Corporation FGFR2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50224
Encompass_Preferred
FGFR2 Colorimetric Cell-Based ELISA Kit for detecting FGFR2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P21802. Gene ID: 2263. Target Synonyms: BEK; BFR-1; ECT1; FGFR-2; Fibroblast growth factor receptor 2 precursor; Keratinocyte growth factor receptor; kinase FGFR2; KSAM-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6942