Learn More
Biomatik Corporation GANP Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50245
Encompass_Preferred
GANP Colorimetric Cell-Based ELISA Kit for detecting GANP in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O60318. Gene ID: 8888. Target Synonyms: 80 kda MCM3-associated protein; GANP protein; KIAA0572; MAP80; MCM3A; MCM3AP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6959