Learn More
Biomatik Corporation GRB10 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50261
Encompass_Preferred
GRB10 Colorimetric Cell-Based ELISA Kit for detecting GRB10 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13322. Gene ID: 2887. Target Synonyms: GRB10 adaptor protein; GRBIR; Growth factor receptor-bound protein 10; Insulin receptor binding protein GRB-IR; KIAA0207. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6973