Learn More
Biomatik Corporation Guanylate Cyclase β Colorimetric Cell-Based ELISA Kit
Catalog No. 502296979
Encompass_Preferred
Guanylate Cyclase β Colorimetric Cell-Based ELISA Kit for detecting Guanylate Cyclase β in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q02153. Gene ID: 2983. Target Synonyms: GCS-beta-1; GCS-beta-3; GCYB1; guanylate cyclase soluble subunit beta-1; GUCY1B1; soluble guanylate cyclase small subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-6979 Supplier Biomatik Corporation Supplier No. EKA50267
Guanylate Cyclase β Colorimetric Cell-Based ELISA Kit for detecting Guanylate Cyclase β in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q02153. Gene ID: 2983. Target Synonyms: GCS-beta-1; GCS-beta-3; GCYB1; guanylate cyclase soluble subunit beta-1; GUCY1B1; soluble guanylate cyclase small subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.