Learn More
Biomatik Corporation HDAC2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50273
Encompass_Preferred
HDAC2 Colorimetric Cell-Based ELISA Kit for detecting HDAC2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q92769. Gene ID: 3066. Target Synonyms: HD2; histone deacetylase 2; YY1 transcription factor binding protein; YY1BP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6985