Learn More
Biomatik Corporation HGS Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50283
Encompass_Preferred
HGS Colorimetric Cell-Based ELISA Kit for detecting HGS in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O14964. Gene ID: 9146. Target Synonyms: Hepatocyte growth factor-regulated tyrosine kinase substrate; HGS; IMOS-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6994