Learn More
Biomatik Corporation JAK3 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50330
Encompass_Preferred
JAK3 Colorimetric Cell-Based ELISA Kit for detecting JAK3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P52333. Gene ID: 3718. Target Synonyms: JAK-3; JAK3; Janus kinase 3; L-JAK; Leukocyte janus kinase; kinase Jak3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-7034