Learn More
Biomatik Corporation Mouse COL3 (Collagen Type III) ELISA Kit
Supplier:  Biomatik Corporation EKF59063
Encompass_Preferred
Mouse COL3 (Collagen Type III) ELISA Kit for detecting COL3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08121. Target Synonyms: COL3; COL3α1; COL3A1; EDS4A; COL3A1; EDS4A; alpha1; III collagen; Collagen 3; collagen alpha-1; III chain; collagen; fetal; collagen; type III; alpha 1; Collagen-3; EDS4A; Ehlers-Danlos syndrome type IV; autosomal dominant. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6268