Learn More
Biomatik Corporation Mouse GS (Gelsolin) ELISA Kit
Supplier:  Biomatik Corporation EKF59070
Encompass_Preferred
Mouse GS (Gelsolin) ELISA Kit for detecting GS in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.813-500ng/ml. Sensitivity: 4.688ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P13020. Target Synonyms: GS; Gelsolin; GSN; ADF; AGEL; Brevin; Actin-depolymerizing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6275