Learn More
Biomatik Corporation Human ZAP70 (Tyrosine-protein kinase ZAP-70) ELISA Kit
Supplier:  Biomatik Corporation EKF59074
Encompass_Preferred
Human ZAP70 (Tyrosine-protein kinase ZAP-70) ELISA Kit for detecting ZAP70 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P43403. Target Synonyms: ZAP70; mrtle; mur; SRK; STD; TZK; 70 kDa zeta-associated protein; EC 2.7.10; EC 2.7.10.2; SRKFLJ17679; STDFLJ17670; Syk-related tyrosine kinase; tyrosine-protein kinase ZAP-70; TZK; ZAP-70; zeta-chain; TCR associated protein kinase; 70 kD; zeta-chain; TCR associated protein kinase 70kDa; zeta-chain associated protein kinase; 70kD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6279