Learn More
Biomatik Corporation Mouse GAS6 (Growth Arrest Specific Protein 6) ELISA Kit
Supplier:  Biomatik Corporation EKF59084
Encompass_Preferred
Mouse GAS6 (Growth Arrest Specific Protein 6) ELISA Kit for detecting GAS6 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q61592. Target Synonyms: GAS6; Growth arrest-specific protein 6; AXLLG; AXLLGAXL stimulatory factor; AXSFAXL receptor tyrosine kinase ligand; GAS-6; growth arrest-specific 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6287