Learn More
Biomatik Corporation Rat Xdh (Xanthine dehydrogenase/oxidase) ELISA Kit
Supplier:  Biomatik Corporation EKF59092
Encompass_Preferred
Rat Xdh (Xanthine dehydrogenase/oxidase) ELISA Kit for detecting Xdh in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P22985. Target Synonyms: Xdh. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6294