Learn More
Biomatik Corporation Human S100A1/S100 (Protein S100-A1) ELISA Kit
Supplier:  Biomatik Corporation EKF59101
Encompass_Preferred
Human S100A1/S100 (Protein S100-A1) ELISA Kit for detecting S100A1; S100 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P23297. Target Synonyms: S100A1; S100A; S100 calcium-binding protein A1; protein S100-A1; S100 alpha; S100 calcium-binding protein A1S100; S-100 protein alpha chain; S-100 protein subunit alpha; S100 protein; alpha polypeptide; S100-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6302