Learn More
Biomatik Corporation Rat Thrombomodulin (Thrombomodulin) ELISA Kit
Supplier:  Biomatik Corporation EKF59106
Encompass_Preferred
Rat Thrombomodulin (Thrombomodulin) ELISA Kit for detecting Thrombomodulin in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 93.75-6000pg/ml. Sensitivity: 56.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O35370. Target Synonyms: Thrombomodulin; THBD; AHUS6; BDCA3; CD141; THPH12; THRM; BDCA-3; Fetomodulin; CD141 antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6307