Learn More
Biomatik Corporation Human PRKCSH (Glucosidase 2 subunit beta) ELISA Kit
Supplier:  Biomatik Corporation EKF59143
Encompass_Preferred
Human PRKCSH (Glucosidase 2 subunit beta) ELISA Kit for detecting PRKCSH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P14314. Target Synonyms: PRKCSH; G19P1; PKCSH; Protein kinase C substRate 60.1 kDa protein heavy chain; 80K-H protein; Glucosidase II subunit beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6339