Learn More
Biomatik Corporation Human ACTN2 (Actinin Alpha 2) ELISA Kit
Supplier:  Biomatik Corporation EKF59154
Encompass_Preferred
Human ACTN2 (Actinin Alpha 2) ELISA Kit for detecting ACTN2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P35609. Target Synonyms: ACTN2; Actinin Alpha 2; CMD1AA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6349