Learn More
Biomatik Corporation Human ITGA2B (Integrin alpha-IIb) ELISA Kit
Supplier:  Biomatik Corporation EKF59159
Encompass_Preferred
Human ITGA2B (Integrin alpha-IIb) ELISA Kit for detecting ITGA2B in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08514. Target Synonyms: ITGA2B; CD41; Integrin alpha-Iib; GP2B; GPIIb; GTA; HPA3; CD41 antigen; GPalpha IIb; ITGAB; platelet fibrinogen receptor; alpha subunit; Platelet membrane glycoprotein IIb; platelet-specific antigen BAK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6352