Learn More
Biomatik Corporation Human TNFRSF17 (Tumor necrosis factor receptor superfamily member 17) ELISA Kit
Supplier:  Biomatik Corporation EKF59161
Encompass_Preferred
Human TNFRSF17 (Tumor necrosis factor receptor superfamily member 17) ELISA Kit for detecting TNFRSF17 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q02223. Target Synonyms: B cell matuRation protein; BCM; BCMA; CD269; TNFRSF17. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6354