Learn More
Biomatik Corporation Mouse Cdh11 (Cadherin-11) ELISA Kit
Supplier:  Biomatik Corporation EKF59180
Encompass_Preferred
Mouse Cdh11 (Cadherin-11) ELISA Kit for detecting Cdh11 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P55288. Target Synonyms: Cdh11; Cadherin-11; CDHOB; OB-Cadherin; CAD11; OSF-4; OB; Osteoblast cadherin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6367