Learn More
Biomatik Corporation Mouse FBXO32 (F-Box Protein 32) ELISA Kit
Supplier:  Biomatik Corporation EKF59182
Encompass_Preferred
Mouse FBXO32 (F-Box Protein 32) ELISA Kit for detecting FBXO32 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 39.063-2500pg/ml. Sensitivity: 23.438pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9CPU7. Target Synonyms: FBXO32. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6369