Learn More
Biomatik Corporation Rat Myosin heavy chain ELISA Kit
Supplier:  Biomatik Corporation EKF59196
Encompass_Preferred
Rat Myosin heavy chain ELISA Kit for detecting myosin heavy chain in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12847. Target Synonyms: Bsh; CG17927; Dm II; Dmel\CG17927; DmMHC; DROMHC; DroMII; FBgn0002741; Ifm; 22; l; 236Ae; l; 2k10423; l; 2M66; Mhc; Mhc36B; mMHC; MRP; Myo; Myosin heavy chain; Nup; Sht; sMHC; Stp. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6383