Learn More
Biomatik Corporation Mouse TLR1 (Toll-like receptor 1) ELISA Kit
Supplier:  Biomatik Corporation EKF59200
Encompass_Preferred
Mouse TLR1 (Toll-like receptor 1) ELISA Kit for detecting TLR1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9EPQ1. Target Synonyms: TLR1; CD281; CD281; CD281 antigen; DKFZp547I0610; DKFZp564I0682; KIAA0012MGC126311; MGC104956; MGC126312; rsc786; TIL; TIL. LPRS5; Toll; interleukin-1 receptor-like protein; toll-like receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6386