Learn More
Biomatik Corporation Human ABCB4 (Multidrug resistance protein 3) ELISA Kit
Supplier:  Biomatik Corporation EKF59202
Encompass_Preferred
Human ABCB4 (Multidrug resistance protein 3) ELISA Kit for detecting ABCB4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P21439. Target Synonyms: ABCB4; Multidrug resistance protein 3; MDR3; PGY3; ATP-binding cassette sub-family B member 4; P-glycoprotein 3; ABC21; GBD1; PFIC-3; ABC21; MDR2; MDR3; MDR2; 3; multidrug resistance protein 3; multiple drug resistance 3; P glycoprotein 3; multiple drug resistance 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6388