Learn More
Biomatik Corporation Human IRF5 (Interferon Regulatory Factor 5) ELISA Kit
Supplier:  Biomatik Corporation EKF59209
Encompass_Preferred
Human IRF5 (Interferon Regulatory Factor 5) ELISA Kit for detecting IRF5 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q13568. Target Synonyms: IRF5; Interferon regulatory factor 5; IRF-5; IBD14; interferon regulatory factor 5; IRF-5; SLEB10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6393