Learn More
Biomatik Corporation Bovine LH (Luteinizing Hormone) ELISA Kit
Supplier:  Biomatik Corporation EKF59234
Encompass_Preferred
Bovine LH (Luteinizing Hormone) ELISA Kit for detecting LH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 1.563-100miu/ml. Sensitivity: 0.938miu/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P04651. Target Synonyms: LH; Luteinizing Hormone; Lutropin; Lutrophin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6405