Learn More
Biomatik Corporation Human USP18 (Ubiquitin Specific Peptidase18) ELISA Kit
Supplier:  Biomatik Corporation EKF59493
Encompass_Preferred
Human USP18 (Ubiquitin Specific Peptidase18) ELISA Kit for detecting USP18 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9UMW8. Target Synonyms: Ubiquitin Specific Peptidase18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6413