Learn More
Biomatik Corporation Mouse Iapp (Islet amyloid polypeptide) ELISA Kit
Supplier:  Biomatik Corporation EKF59629
Encompass_Preferred
Mouse Iapp (Islet amyloid polypeptide) ELISA Kit for detecting Iapp in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12968. Target Synonyms: IAPP; DAP; IAP; Amylin; Islet amyloid polypeptide; Insulinoma amyloid peptide; Diabetes-associated peptide; DAP;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6420