Learn More
Biomatik Corporation Porcine PGE2 (Prostaglandin E2) ELISA Kit
Supplier:  Biomatik Corporation EKF59698
Encompass_Preferred
Porcine PGE2 (Prostaglandin E2) ELISA Kit for detecting PGE2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: Prostaglandin E2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6422