Learn More
Biomatik Corporation Rat CXCR4 (C-X-C motif receptor 4) ELISA Kit
Supplier:  Biomatik Corporation EKF59752
Encompass_Preferred
Rat CXCR4 (C-X-C motif receptor 4) ELISA Kit for detecting CXCR4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O08565. Target Synonyms: CXCR4; Chemokine C-X-C-Motif Receptor; Stromal cell-derived factor 1 receptor; SDF-1 receptor; Leukocyte-derived seven transmembrane domain receptor; LESTR; Fusin; LCR1; CD184; Fusin; chemokine; C-X-C motif receptor 4; C-X-C chemokine receptor type 4; CXC-R4; ; D2S201ESDF-1 receptor; FB22; Fusin; HM89NPYRL; HSY3RR; LAP3; LCR1; LESTRCD184 antigen; Leukocyte-derived seven transmembrane domain receptor; lipopolysaccharide-associated protein 3; neuropeptide Y receptor Y3; NPY3RWHIM; NPYR; NPYY3R. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6429