Learn More
Biomatik Corporation ET-1 (Endothelin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF60092
Encompass_Preferred
ET-1 (Endothelin-1) ELISA Kit for detecting ET-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 1.25-80pg/ml. Sensitivity: 0.75pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P05305. Target Synonyms: ET-1; EDN1; ET1; endothelin 1; endothelin-1; preproendothelin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6448