Learn More
Biomatik Corporation Mouse IL-1ra (Interleukin 1 Receptor Antagonist) ELISA Kit
Supplier:  Biomatik Corporation EKF60119
Encompass_Preferred
Mouse IL-1ra (Interleukin 1 Receptor Antagonist) ELISA Kit for detecting IL-1ra in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P25085. Target Synonyms: IL-1RA; IL-1ra; IL-1F3; IL1RN; DIRA; ICIL-1RA; IL-1RN; IL-1ra3; IL1F3; IL1RA; IRAP; MVCD4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6450