Learn More
Biomatik Corporation Human TMEM173 (Transmembrane protein 173) ELISA Kit
Supplier:  Biomatik Corporation EKF60255
Encompass_Preferred
Human TMEM173 (Transmembrane protein 173) ELISA Kit for detecting TMEM173 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q86WV6. Target Synonyms: hMITA; hSTING; Mediator of IRF3 activation; MITA; MPYS; NET23; STING; TMEM173; transmembrane protein 173. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6457