Learn More
Biomatik Corporation Human WTIP (Wilms tumor protein 1-interacting protein) ELISA Kit
Supplier:  Biomatik Corporation EKF60394
Encompass_Preferred
Human WTIP (Wilms tumor protein 1-interacting protein) ELISA Kit for detecting WTIP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: A6NIX2. Target Synonyms: WTIP; Wilms tumor protein 1-interacting protein; WT1-interacting protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6463