Learn More
Biomatik Corporation Human TMPRSS6 (Transmembrane protease serine 6) ELISA Kit
Supplier:  Biomatik Corporation EKF60410
Encompass_Preferred
Human TMPRSS6 (Transmembrane protease serine 6) ELISA Kit for detecting TMPRSS6 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8IU80. Target Synonyms: IRIDA; Matriptase 2; TMPRSS6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6466