Learn More
Biomatik Corporation Human PDI (Protein Disulfide Isomerase) ELISA Kit
Supplier:  Biomatik Corporation EKF60416
Encompass_Preferred
Human PDI (Protein Disulfide Isomerase) ELISA Kit for detecting PDI in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07237. Target Synonyms: PDI; DSI; ERBA2L; GIT; PDIA1; PO4DB; PO4HB; Thbp; Cellular thyroid hormone-binding protein; collagen prolyl 4-hydroxylase beta; DSI; ERBA2L; GIT; glutathione-insulin transhydrogenase; P4Hbeta; p55; PDIA1procollagen-proline; 2-oxoglutaRate 4-dioxygenase; proline 4-hydroxylase; betapolypeptide; protein disulfide isomerase-associated 1; PHDB; procollagen-proline; 2-oxoglutaRate 4-dioxygenase; proline 4-hydroxylase; betapolypeptide; PROHBprocollagen-proline; 2-oxoglutaRate 4-dioxygenase; proline 4-hydroxylase; betapolypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6470