Learn More
Biomatik Corporation Mouse Ckm (Creatine kinase M-type) ELISA Kit
Supplier:  Biomatik Corporation EKF60425
Encompass_Preferred
Mouse Ckm (Creatine kinase M-type) ELISA Kit for detecting Ckm in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07310. Target Synonyms: Ckm; CKMM; M-CK; Creatine kinase M chain; creatine kinase; muscle; creatine kinase-M. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6475