Learn More
Biomatik Corporation Human ANXA2 (Annexin A2) ELISA Kit
Supplier:  Biomatik Corporation EKF60432
Encompass_Preferred
Human ANXA2 (Annexin A2) ELISA Kit for detecting ANXA2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07355. Target Synonyms: ANXA2; Annexin A2; Lipocortin II; Chromobindin-8; Calpactin-1 heavy chain; Calpactin I heavy chain; p36; Protein I; Placental anticoagulant protein IV; PAP-IV; Annexin II; Annexin-2; ANX2; ANX2L4; CAL1H; LPC2D; LIP2; LPC2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6480