Learn More
Biomatik Corporation Human TLR9 (Toll-like receptor 9) ELISA Kit
Supplier:  Biomatik Corporation EKF60435
Encompass_Preferred
Human TLR9 (Toll-like receptor 9) ELISA Kit for detecting TLR9 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9NR96. Target Synonyms: TLR9; CD289; Toll-like receptor 9; CD289 antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6482