Learn More
Biomatik Corporation Human CYP1A2 (Cytochrome P450, family 1, subfamily A, polypeptide 2) ELISA Kit
Supplier:  Biomatik Corporation EKF60451
Encompass_Preferred
Human CYP1A2 (Cytochrome P450, family 1, subfamily A, polypeptide 2) ELISA Kit for detecting CYP1A2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P05177. Target Synonyms: CYP1A2; Cytochrome P450 1A2; Cytochrome P450-P3; Cytochrome P450 4; CYPIA2; Cytochrome P; 3450. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6490