Learn More
Biomatik Corporation Human LARP7 (La-related protein 7) ELISA Kit
Supplier:  Biomatik Corporation EKF60461
Encompass_Preferred
Human LARP7 (La-related protein 7) ELISA Kit for detecting LARP7 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q4G0J3. Target Synonyms: LARP7; La-related protein 7; La ribonucleoprotein domain family member 7; P-TEFb-interaction protein for 7SK stability; PIP7S; HDCMA18P. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6496